missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(methylamino)piperidine

Catalog No. AAH50985MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50985MD 250 mg
AAH5098503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50985MD Supplier Thermo Scientific Chemicals Supplier No. H50985MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139062-96-7 | C8H16N2O | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139062-96-7
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.23
MDL Number MFCD07367773
InChI Key RSEPODZAQBVPOS-UHFFFAOYSA-N
Synonym 1-acetyl-4-methylamino piperidine, 1-acetyl-n-methylpiperidin-4-amine, 1-4-methylamino piperidin-1-yl ethan-1-one, 1-4-methylamino piperidin-1-yl ethanone, 4-n-methylamino-1-acetyl-piperidine, 4-piperidinamine, 1-acetyl-n-methyl-9ci, 4-n-methylamino-1-acetyl, acmc-209vt3, 1-acetyl-4-methylaminopiperidine
PubChem CID 19761265
SMILES CNC1CCN(CC1)C(C)=O

Specifications

Quantity 250 mg
UN Number UN3259
IUPAC Name 1-[4-(methylamino)piperidin-1-yl]ethan-1-one
Formula Weight 156.23
Chemical Name or Material 1-Acetyl-4-(methylamino)piperidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only