missing translation for 'onlineSavingsMsg'
Learn More

1-Acenaphthenol 97.0+%, TCI America™

Catalog No. A10575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10575G Supplier TCI America Supplier No. A10575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Organic building block used in chemical research and synthesis.

Chemical Identifiers

CAS 6306-07-6
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00003808
InChI Key MXUCIEHYJYRTLT-UHFFFAOYNA-N
Synonym 1-acenaphthenol, acenaphthen-1-ol, acenaphthenol, 7-acenaphthenol, acenaphthenol-1, 1-hydroxyacenaphthene, 1-acenaphthylenol, 1,2-dihydro, acenaphthene-1-ol, acenaphthylenol, 1,2-dihydro, 1-acenaphthalenol
PubChem CID 22750
IUPAC Name 1,2-dihydroacenaphthylen-1-ol
SMILES C1C(C2=CC=CC3=C2C1=CC=C3)O

Specifications

Melting Point 146°C
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acenaphthenol