missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanedithiol, 95%

Catalog No. AAL0717203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0717203 1 g
AAL0717206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0717203 Supplier Thermo Scientific Chemicals Supplier No. L0717203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3489-28-9 | C9H20S2 | 192.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3489-28-9
Molecular Formula C9H20S2
Molecular Weight (g/mol) 192.38
MDL Number MFCD00004914
InChI Key GJRCLMJHPWCJEI-UHFFFAOYSA-N
Synonym 1,9-nonanedithiol, 1,9-dimercaptononane, nonamethylene dimercaptan, unii-5bpj1xqe3t, 5bpj1xqe3t, fema no. 3513, 1,9-nonane dithiol, 1,9-dinonyl mercaptan, acmc-20ap0g
PubChem CID 248488
IUPAC Name nonane-1,9-dithiol
SMILES SCCCCCCCCCS

Specifications

Density 0.953
Boiling Point 351°C to 352°C
Flash Point >100°C (212°F)
Refractive Index 1.4999
Quantity 1 g
Beilstein 1737568
Sensitivity Air Sensitive
Formula Weight 192.39
Percent Purity 97%
Chemical Name or Material 1,9-Nonanedithiol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 222-482-3
TSCA Yes
Recommended Storage Store under Argon

RUO – Research Use Only