missing translation for 'onlineSavingsMsg'
Learn More

1,9-Bis(acryloyloxy)nonane (stabilized with MEHQ) 92.0+%, TCI America™

Catalog No. p-7119811
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B286325G 25 g
B2863500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B286325G Supplier TCI America Supplier No. B286325G
Only null left

Chemical Identifiers

CAS 107481-28-7
Molecular Formula C15H24O4
Molecular Weight (g/mol) 268.35
MDL Number MFCD00080592
InChI Key PGDIJTMOHORACQ-UHFFFAOYSA-N
Synonym Nonamethylene Glycol Diacrylate, Nonanediol Diacrylate
PubChem CID 9795378
IUPAC Name 9-(prop-2-enoyloxy)nonyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCCCCOC(=O)C=C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 268.35
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,9-Bis(acryloyloxy)nonane (stabilized with MEHQ)
EINECSNumber (2)-1008
TSCA Yes