missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,9-Bis(acryloyloxy)nonane (stabilized with MEHQ) 92.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 268.35 |
| Percent Purity | ≥92.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,9-Bis(acryloyloxy)nonane (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-1008 |
| TSCA | Yes |