missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanediol 99.0+%, TCI America™

Catalog No. O002425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002425G 25 g
O0024100G 100 g
O0024500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O002425G Supplier TCI America Supplier No. O002425G

Chemical Identifiers

CAS 629-41-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00002989
InChI Key OEIJHBUUFURJLI-UHFFFAOYSA-N
PubChem CID 69420
ChEBI CHEBI:44630
IUPAC Name octane-1,8-diol
SMILES C(CCCCO)CCCO

Specifications

Melting Point 62°C
Color White
Boiling Point 166°C
Quantity 25 g
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Formula Weight 146.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Octanediol
EINECSNumber (2)-0240
TSCA No