missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Octanediol 99.0+%, TCI America™
Supplier: TCI America O0024100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| 629-41-4 | |
| 146.23 | |
| OEIJHBUUFURJLI-UHFFFAOYSA-N | |
| CHEBI:44630 | |
| C(CCCCO)CCCO |
| C8H18O2 | |
| MFCD00002989 | |
| 69420 | |
| octane-1,8-diol |
Specifications
| 1,8-Octanediol | |
| 62°C | |
| 166°C | |
| 100 g | |
| 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p | |
| C(CCCCO)CCCO | |
| 146.23 | |
| CHEBI:44630 | |
| ≥99.0% (GC) |
| 629-41-4 | |
| White | |
| C8H18O2 | |
| MFCD00002989 | |
| OEIJHBUUFURJLI-UHFFFAOYSA-N | |
| octane-1,8-diol | |
| 69420 | |
| 146.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : No