missing translation for 'onlineSavingsMsg'
Learn More

1,8-Nonadiyne 97.0+%, TCI America™

Catalog No. N04065ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04065ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N04065ML Supplier TCI America Supplier No. N04065ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2396-65-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008581
InChI Key DMOVPHYFYSASTC-UHFFFAOYSA-N
Synonym 1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 16969
IUPAC Name nona-1,8-diyne
SMILES C#CCCCCCC#C

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Nonadiyne
TSCA Yes
Recommended Storage Refrigerator