missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Naphthalimide 98.0+%, TCI America™
$407.67 - $407.67
Chemical Identifiers
| CAS | 81-83-4 |
|---|---|
| Molecular Formula | C12H7NO2 |
| Molecular Weight (g/mol) | 197.193 |
| MDL Number | MFCD00006920 |
| InChI Key | XJHABGPPCLHLLV-UHFFFAOYSA-N |
| Synonym | 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn |
| PubChem CID | 66491 |
| IUPAC Name | benzo[de]isoquinoline-1,3-dione |
| SMILES | C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2 |
Chemical Identifiers
| 81-83-4 | |
| 197.193 | |
| XJHABGPPCLHLLV-UHFFFAOYSA-N | |
| 66491 | |
| C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2 |
| C12H7NO2 | |
| MFCD00006920 | |
| 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn | |
| benzo[de]isoquinoline-1,3-dione |
Specifications
| 81-83-4 | |
| C12H7NO2 | |
| 250 g | |
| XJHABGPPCLHLLV-UHFFFAOYSA-N | |
| benzo[de]isoquinoline-1,3-dione | |
| 66491 | |
| ≥98.0% (HPLC,N) | |
| 1,8-Naphthalimide |
| 301°C | |
| MFCD00006920 | |
| 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn | |
| C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2 | |
| 197.193 | |
| 197.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes