missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalimide 98.0+%, TCI America™

Catalog No. N0456250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0456250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0456250G Supplier TCI America Supplier No. N0456250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-83-4
Molecular Formula C12H7NO2
Molecular Weight (g/mol) 197.193
MDL Number MFCD00006920
InChI Key XJHABGPPCLHLLV-UHFFFAOYSA-N
Synonym 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn
PubChem CID 66491
IUPAC Name benzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2

Specifications

Melting Point 301°C
Quantity 250 g
Formula Weight 197.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Naphthalimide