missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalimide, 98%

Catalog No. p-7024352
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1885609 10 g
AAA1885618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1885609 Supplier Thermo Scientific Chemicals Supplier No. A1885609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 81-83-4 | C12H7NO2 | 197.193 g/mol

It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 81-83-4
Molecular Formula C12H7NO2
Molecular Weight (g/mol) 197.193
MDL Number MFCD00006920
InChI Key XJHABGPPCLHLLV-UHFFFAOYSA-N
Synonym 1,8-naphthalimide, naphthalimide, 1h-benz de isoquinoline-1,3 2h-dione, 1h-benzo de isoquinoline-1,3 2h-dione, 1,8-naphthalenedicarboximide, benzo de isoquinoline-1,3-dione, unii-gi0tv19gbn, naphthalene-1,8-dicarboximide, gi0tv19gbn
PubChem CID 66491
IUPAC Name benzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2

Specifications

Melting Point 299°C to 303°C
Quantity 10 g
Beilstein 153150
Solubility Information Insoluble in water.
Formula Weight 197.2
Percent Purity 98%
Chemical Name or Material 1,8-Naphthalimide
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-379-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only