missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, 95%

CAS: 117-10-2 | C14H8O4 | 240.21 g/mol

$59.31 - $59.31

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001211
InChI Key QBPFLULOKWLNNW-UHFFFAOYSA-N
Synonym danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
PubChem CID 2950
ChEBI CHEBI:3682
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114830050
SDS View Documents
Thermo Scientific Chemicals
114830050
5 g Glass bottle
Each for $59.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

117-10-2
240.21
QBPFLULOKWLNNW-UHFFFAOYSA-N
2950
1,8-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001211
danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
CHEBI:3682
C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O

Specifications

117-10-2
100.0
Brown or Brown-Orange
Authentic
Glass bottle
MFCD00001211
08, 458
danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
QBPFLULOKWLNNW-UHFFFAOYSA-N
1,8-dihydroxyanthracene-9,10-dione
2950
240.21
Powder
90.0
190.0°C to 195.0°C
>200°C
94% min. (HPLC)
C14H8O4
5 g
15, 2814
Solubility in water: insoluble. Other solubilities: almost insoluble in alcohol,moderately soluble in ether,chloroform,soluble in 10 parts hot glacial acetic acid
C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O
240.21
CHEBI:3682
95%
1, 8-Dihydroxyanthraquinone, 90-95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes serious eye irritation.

GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 204-173-5

RTECSNumber : CB6650000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only