missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, 95%

Catalog No. AC114830050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114830050 5 g Glass bottle
AC114831000 100 g Glass bottle
AC114835000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114830050 Supplier Thermo Scientific Chemicals Supplier No. 114830050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 117-10-2 | C14H8O4 | 240.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001211
InChI Key QBPFLULOKWLNNW-UHFFFAOYSA-N
Synonym danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
PubChem CID 2950
ChEBI CHEBI:3682
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color Brown or Brown-Orange
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 458
Merck Index 15, 2814
Solubility Information Solubility in water: insoluble. Other solubilities: almost insoluble in alcohol,moderately soluble in ether,chloroform,soluble in 10 parts hot glacial acetic acid
Formula Weight 240.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1, 8-Dihydroxyanthraquinone, 90-95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 204-173-5
RTECSNumber CB6650000
TSCA TSCA

RUO – Research Use Only