missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dichlorooctane 99.0+%, TCI America™
$87.60 - $147.25
Chemical Identifiers
| CAS | 2162-99-4 |
|---|---|
| Molecular Formula | C8H16Cl2 |
| Molecular Weight (g/mol) | 183.12 |
| MDL Number | MFCD00039393 |
| InChI Key | WXYMNDFVLNUAIA-UHFFFAOYSA-N |
| Synonym | octane, 1,8-dichloro, 1,8-dichloroethane, octamethylene chloride, octamethylene dichloride, acmc-209fmk, 1,8-dichlorooctane |
| PubChem CID | 75102 |
| IUPAC Name | 1,8-dichlorooctane |
| SMILES | ClCCCCCCCCCl |
Chemical Identifiers
| 2162-99-4 | |
| 183.12 | |
| WXYMNDFVLNUAIA-UHFFFAOYSA-N | |
| 75102 | |
| ClCCCCCCCCCl |
| C8H16Cl2 | |
| MFCD00039393 | |
| octane, 1,8-dichloro, 1,8-dichloroethane, octamethylene chloride, octamethylene dichloride, acmc-209fmk, 1,8-dichlorooctane | |
| 1,8-dichlorooctane |
Specifications
| 2162-99-4 | |
| 116°C | |
| MFCD00039393 | |
| 3082 | |
| WXYMNDFVLNUAIA-UHFFFAOYSA-N | |
| 1,8-dichlorooctane | |
| 75102 | |
| ≥99.0% (GC) | |
| 1,8-Dichlorooctane |
| Yellow | |
| C8H16Cl2 | |
| 10 g | |
| octane, 1,8-dichloro, 1,8-dichloroethane, octamethylene chloride, octamethylene dichloride, acmc-209fmk, 1,8-dichlorooctane | |
| ClCCCCCCCCCl | |
| 183.12 | |
| 183.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0066
TSCA : Yes