missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dichloroanthraquinone 95.0+%, TCI America™

Catalog No. p-7126149
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032925G 25 g
D0329250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D032925G Supplier TCI America Supplier No. D032925G
Only null left

Chemical Identifiers

CAS 82-43-9
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001191
InChI Key VBQNYYXVDQUKIU-UHFFFAOYSA-N
Synonym 1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
PubChem CID 6708
IUPAC Name 1,8-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 25 g
Formula Weight 277.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Dichloroanthraquinone
EINECSNumber (4)-0689
RTECSNumber CB6496000
TSCA Yes