missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dichloroanthraquinone, 96%

Catalog No. p-7026999
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2342114 25 g
AAB2342122 100 g
AAB2342136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2342114 Supplier Thermo Scientific Chemicals Supplier No. B2342114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82-43-9 | C14H6Cl2O2 | 277.1 g/mol

1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 82-43-9
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001191
InChI Key VBQNYYXVDQUKIU-UHFFFAOYSA-N
Synonym 1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
PubChem CID 6708
IUPAC Name 1,8-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl

Specifications

Melting Point 201°C to 203°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 277.11
Percent Purity 96%
Chemical Name or Material 1,8-Dichloroanthraquinone
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 201-420-9
RTECSNumber CB6496000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only