missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromonaphthalene 98.0+%, TCI America™

Catalog No. D46561G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D46561G 1 g
D46565G 5 g
2 options

Catalog No. D46561G

Supplier: TCI America D46561G

Only null left

Chemical Identifiers

CAS 17135-74-9
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.966
MDL Number MFCD00183574
InChI Key DLXBGTIGAIESIG-UHFFFAOYSA-N
PubChem CID 625356
IUPAC Name 1,8-dibromonaphthalene
SMILES C1=CC2=C(C(=C1)Br)C(=CC=C2)Br

Specifications

Melting Point 110°C
Boiling Point 140°C
Quantity 1 g
Formula Weight 285.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Dibromonaphthalene