missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%

Catalog No. p-7082155
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1244914 25 g
AAA1244922 100 g
AAA1244936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1244914 Supplier Thermo Scientific Chemicals Supplier No. A1244914
Only null left

CAS: 6674-22-2 | C9H16N2 | 152.241 g/mol

1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.

Chemical Identifiers

CAS 6674-22-2
Molecular Formula C9H16N2
Molecular Weight (g/mol) 152.241
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

Melting Point -70°C
Density 1.02
pH 12.8
Boiling Point 261°C
Flash Point 116°C (240°F)
Odor Amine-like
Refractive Index 1.522
Quantity 25 g
UN Number UN3267
Beilstein 508906
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 229-713-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only