missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]-7-undecene 98.0+%, TCI America™

Catalog No. D1270500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1270500G 500 g
D127025G 25 g
D1270100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1270500G Supplier TCI America Supplier No. D1270500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6674-22-2
Molecular Formula C9H16N2
Molecular Weight (g/mol) 152.241
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

Melting Point -70°C
Color Yellow
pH 12.8 (10g/L H2O soln. 20°C)
Boiling Point 83°C
Quantity 500 g
UN Number 3267
Formula Weight 152.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]-7-undecene
EINECSNumber (5)-1117
TSCA Yes