missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diacetoxyoctane, TCI America™

Catalog No. O027525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O027525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O027525G Supplier TCI America Supplier No. O027525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3992-48-1
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD01861294
InChI Key MDHIGESKEUQAEO-UHFFFAOYSA-N
Synonym Octamethylene Diacetate, 1,8-Octanediol Diacetate
PubChem CID 5216131
IUPAC Name 8-acetyloxyoctyl acetate
SMILES CC(=O)OCCCCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 g
Formula Weight 230.30
Physical Form Liquid
Chemical Name or Material 1,8-Diacetoxyoctane