missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiene 97.0+%, TCI America™

Catalog No. O010425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O010425ML 25 mL
O0104100ML 100 mL
O0104500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O010425ML Supplier TCI America Supplier No. O010425ML

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 2309
Formula Weight 110.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Octadiene
EINECSNumber (2)-3935
RTECSNumber RG5250000
TSCA Yes