missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiene Diepoxide 97.0+%, TCI America™

Catalog No. p-7126805
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15865G 5 g
D158625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15865G Supplier TCI America Supplier No. D15865G
Only null left

Chemical Identifiers

CAS 2426-07-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00005155
InChI Key LFKLPJRVSHJZPL-UHFFFAOYNA-N
Synonym 1,2,7,8-diepoxyoctane, 1,2:7,8-diepoxyoctane, 1,7-octadiene diepoxide, oxirane, 2,2'-1,4-butanediyl bis, 1,2-epoxy-7,8-epoxyoctane, 2,2'-1,4-butanediyl bisoxirane, 1,4-di oxiran-2-yl butane, octane, 1,2:7,8-diepoxy, 2-4-oxiran-2-yl butyl oxirane, ccris 963
PubChem CID 17048
ChEBI CHEBI:23705
IUPAC Name 2-[4-(oxiran-2-yl)butyl]oxirane
SMILES C1C(O1)CCCCC2CO2

Specifications

Color Yellow
Boiling Point 101°C
Quantity 5 g
UN Number 2810
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Octadiene Diepoxide
RTECSNumber RG9450000
TSCA No