missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,7-Octadiene, 98.5%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Melting Point | -70.0°C |
| Color | Colorless to Yellow |
| Density | 0.7400g/mL |
| Boiling Point | 114.0°C to 121.0°C |
| Flash Point | 9°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.50% |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge |
| EINECSNumber | 223-054-9 |
RUO – Research Use Only