missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiene, 98.5%

Catalog No. AC129355000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC129355000 500 mL
AC129351000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129355000 Supplier Thermo Scientific Chemicals Supplier No. 129355000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3710-30-3 | C8H14 | 110.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 114.0°C to 121.0°C
Flash Point 9°C
Infrared Spectrum Authentic
Assay Percent Range 98.50%
Packaging Glass bottle
Refractive Index 1.4210 to 1.4230
Linear Formula H2C=CH(CH2)4CH=CH2
Quantity 500 mL
UN Number 2309
Specific Gravity 0.74
Solubility Information Solubility in water: 0.08g/L (20°C). Other solubilities: miscible with methanol, cyclohexane, benzene,, isopropanol and petroleumether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 110.2
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1, 7-Octadiene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
EINECSNumber 223-054-9

RUO – Research Use Only