missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiene, 97%

Catalog No. aal07659ae
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL07659AE 100 mL
AAL07659AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL07659AE Supplier Thermo Scientific Chemicals Supplier No. L07659AE
Only null left

CAS: 3710-30-3 | C8H14 | 110.20 g/mol

1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Octadiene is used in organic synthesis. It is also used to study and assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. Further, it is used in a study to investigate micropatterned surfaces prepared by plasma polymerization. It acts as a crosslinker as well as a source of ethylene in cross-enyne metathesis (CEYM)-related reactions.

Solubility
Miscible with methanol and cyclohexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

Melting Point <-70°C
Density 0.735
Boiling Point 118°C to 120°C
Flash Point 5°C (41°F)
Odor Strong
Refractive Index 1.422
Linear Formula CH2=CH(CH2)4CH=CH2
Quantity 100 mL
UN Number UN2309
Beilstein 605288
Solubility Information Miscible with methanol and cyclohexane. Slightly miscible with water.
Formula Weight 110.2
Percent Purity 97%
Chemical Name or Material 1,7-Octadiene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTADIENE
EINECSNumber 223-054-9
RTECSNumber RG5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only