missing translation for 'onlineSavingsMsg'
Learn More

1,7-Heptanediol 98.0+%, TCI America™

Catalog No. H00285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00285G 5 g
H002825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00285G Supplier TCI America Supplier No. H00285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-30-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.203
MDL Number MFCD00002987
InChI Key SXCBDZAEHILGLM-UHFFFAOYSA-N
Synonym 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor
PubChem CID 12381
IUPAC Name heptane-1,7-diol
SMILES C(CCCO)CCCO

Specifications

Color Yellow
Boiling Point 262°C
Quantity 5 g
Formula Weight 132.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Heptanediol
EINECSNumber (2)-0240
RTECSNumber MI9804000
TSCA No