missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,7-Heptanediol 98.0+%, TCI America™
Supplier: TCI America H002825G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C7H16O2 | |
| MFCD00002987 | |
| 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor | |
| heptane-1,7-diol |
Specifications
| 1,7-Heptanediol | |
| Yellow | |
| C7H16O2 | |
| MFCD00002987 | |
| SXCBDZAEHILGLM-UHFFFAOYSA-N | |
| heptane-1,7-diol | |
| 12381 | |
| ≥98.0% (GC) |
| 629-30-1 | |
| 262°C | |
| 25 g | |
| 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor | |
| C(CCCO)CCCO | |
| 132.203 | |
| 132.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0240
RTECSNumber : MI9804000
TSCA : No