missing translation for 'onlineSavingsMsg'
Learn More

1,7-Heptanediol, 95%

Catalog No. AAL0638406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0638406 5 g
AAL0638414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0638406 Supplier Thermo Scientific Chemicals Supplier No. L0638406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-30-1 | C7H16O2 | 132.203 g/mol

1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Heptanediol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water, slightly soluble in ether.

Notes
Incompatible with reducing agents, oxidizing agents, Acid chlorides, Acid anhydrides and chloroformates. Store away from acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 629-30-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.203
MDL Number MFCD00002987
InChI Key SXCBDZAEHILGLM-UHFFFAOYSA-N
Synonym 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor
PubChem CID 12381
IUPAC Name heptane-1,7-diol
SMILES C(CCCO)CCCO

Specifications

Melting Point 17°C to 18°C
Density 0.953
Boiling Point 258°C to 260°C
Flash Point 127°C (260°F)
Refractive Index 1.455
Quantity 5 g
Beilstein 1633482
Solubility Information Soluble in water,slightly soluble in ether.
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 1,7-Heptanediol
Show More Show Less
EINECSNumber 211-085-0
RTECSNumber MI9804000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only