missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dioxaspiro[5.5]undecane, 98%

Catalog No. AAB2166406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2166406 5 g
AAB2166414 25 g
AAB2166422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2166406 Supplier Thermo Scientific Chemicals Supplier No. B2166406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 180-84-7 | C9H16O2 | 156.225 g/mol

Suitable for use in organic farming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for use in organic farming.

Solubility
Soluble in water 1.0g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 180-84-7
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00011578
InChI Key GBBVHDGKDQAEOT-UHFFFAOYSA-N
Synonym 1,7-dioxaspiro 5.5 undecane, olean, unii-khi3qeq4zj, 1,7-dioxaspiro 5,5 undecane, khi3qeq4zj, r-1,7-dioxaspiro 5.5 undecane, s-1,7-dioxaspiro 5.5 undecane, olive-fly ketal, eco-trap
PubChem CID 67437
IUPAC Name 1,7-dioxaspiro[5.5]undecane
SMILES C1CCOC2(C1)CCCCO2

Specifications

Density 1.02
Boiling Point 193°C
Flash Point 63°C (145°F)
Refractive Index 1.464
Quantity 5 g
Beilstein 105997
Solubility Information Soluble in water 1.0g/L.
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material 1,7-Dioxaspiro[5.5]undecane
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 205-870-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only