missing translation for 'onlineSavingsMsg'
Learn More

1,7-Diaminoheptane 98.0+%, TCI America™

Catalog No. p-7126026
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00945G 5 g
D009425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00945G Supplier TCI America Supplier No. D00945G
Only null left

Chemical Identifiers

CAS 646-19-5
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008246
InChI Key PWSKHLMYTZNYKO-UHFFFAOYSA-N
Synonym 1,7-diaminoheptane, 1,7-heptanediamine, heptamethylenediamine, chembl28242, h2n ch2 7nh2, heptan-1,7-diamin, 1,9-diazanonane, acmc-1atsg, 1,7-heptamethylenediamine
PubChem CID 69533
IUPAC Name heptane-1,7-diamine
SMILES NCCCCCCCN

Specifications

Color White
Boiling Point 225°C
Quantity 5 g
UN Number 3259
Formula Weight 130.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,7-Diaminoheptane