missing translation for 'onlineSavingsMsg'
Learn More

1,7-Diaminoheptane, 98%, Thermo Scientific™

Catalog No. AC112200250
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC112200250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112200250 Supplier Thermo Scientific Chemicals Supplier No. 112200250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 646-19-5
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 223°C to 225°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H18N2
Linear Formula H2N(CH2)7NH2
Quantity 25g
MDL Number MFCD00008246
Beilstein 04, 271
Synonym 1,7-diaminoheptane, 1,7-heptanediamine, heptamethylenediamine, chembl28242, h2n ch2 7nh2, heptan-1,7-diamin, 1,9-diazanonane, acmc-1atsg, 1,7-heptamethylenediamine
Solubility Information Solubility in water: soluble
InChI Key PWSKHLMYTZNYKO-UHFFFAOYSA-N
SMILES NCCCCCCCN
IUPAC Name heptane-1,7-diamine
Molecular Weight (g/mol) 130.24
PubChem CID 69533
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 1, 7-Diaminoheptane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 211-468-2
TSCA TSCA

RUO – Research Use Only