missing translation for 'onlineSavingsMsg'
Learn More

1,7-Diaminoheptane, 98%

Catalog No. aal0285314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0285314 25 g
AAL0285306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0285314 Supplier Thermo Scientific Chemicals Supplier No. L0285314
Only null left

CAS: 646-19-5 | C7H18N2 | 130.24 g/mol

1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

Solubility
Soluble in water, ethanol, acetone, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.

Chemical Identifiers

CAS 646-19-5
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008246
InChI Key PWSKHLMYTZNYKO-UHFFFAOYSA-N
Synonym 1,7-diaminoheptane, 1,7-heptanediamine, heptamethylenediamine, chembl28242, h2n ch2 7nh2, heptan-1,7-diamin, 1,9-diazanonane, acmc-1atsg, 1,7-heptamethylenediamine
PubChem CID 69533
IUPAC Name heptane-1,7-diamine
SMILES NCCCCCCCN

Specifications

Melting Point 26°C to 30°C
Density 0.86
Boiling Point 223°C to 225°C
Flash Point 87°C (188°F)
Odor Amine-like
Refractive Index 1.459
Linear Formula H2N(CH2)7NH2
Quantity 25 g
UN Number UN3259
Beilstein 1734159
Sensitivity Air Sensitive
Solubility Information Soluble in water,ethanol,acetone,ether and benzene.
Formula Weight 130.24
Percent Purity 98%
Chemical Name or Material 1,7-Diaminoheptane
Show More Show Less
Hazard Category H302-H314-H335-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 211-468-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only