missing translation for 'onlineSavingsMsg'
Learn More

1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Catalog No. AAH26293MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26293MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26293MD Supplier Thermo Scientific Chemicals Supplier No. H26293MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162148-48-3 | C20H40N4O4 | 400.56 g/mol

Intermediate in the preparation of chelating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in the preparation of chelating agents.

Solubility
Soluble in acetone, acetonitrile, ethyl Acetate, chloroforme, cyclohexane. Insoluble in water.

Notes
Hygroscopic. Store in cool place.

Chemical Identifiers

CAS 162148-48-3
Molecular Formula C20H40N4O4
Molecular Weight (g/mol) 400.56
MDL Number MFCD09263303
InChI Key OERWIIJOLSXGAQ-UHFFFAOYSA-N
Synonym di-tert-butyl 2,2'-1,4,7,10-tetraazacyclododecane-1,7-diyl diacetate, 1,7-bis tert-butoxycarbonylmethyl-1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane-1,7-diacetic acid 1,7-bis 1,1-dimethylethyl ester, tert-butyl 2-7-2-tert-butoxy-2-oxoethyl-1,4,7,10-tetraazacyclododecan-1-yl acetate, 1,4,7,10-tetraazacyclododecane-1,7-bis t-butyl acetate, 1,7-bis carbo-tert-butoxymethyl-1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane-1,7-bis acetic acid tert-butyl ester, 1,4,7,10-tetraazacyclododecane-1,7-diacetic acid, bis 1,1-dimethylethyl ester, tert-butyl 2-7-2-tert-butoxy-2-oxo-ethyl-1,4,7,10-tetrazacyclododec-1-yl acetate, tert-butyl 2-7-2-2-methylpropan-2-yl oxy-2-oxoethyl-1,4,7,10-tetrazacyclododec-1-yl acetate
PubChem CID 393881
IUPAC Name tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
SMILES CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCNCC1

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,acetonitrile,ethyl Acetate,chloroforme,cyclohexane. Insoluble in water.
Formula Weight 400.56
Chemical Name or Material 1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only