missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(6-Methoxy-2-naphthyl)ethanol, 98%
Description
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.
Solubility
Soluble in DMSO, acetone, dichloromethane, ethylacetate. Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.
Solubility
Soluble in DMSO, acetone, dichloromethane, ethylacetate. Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 111°C to 112°C |
| Quantity | 25 g |
| Solubility Information | Soluble in DMSO,acetone,dichloromethane,ethylacetate. Insoluble in water. |
| IUPAC Name | 1-(6-methoxynaphthalen-2-yl)ethanol |
| Formula Weight | 202.25 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(6-Methoxy-2-naphthyl)ethanol |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only