missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanedithiol 97.0+%, TCI America™

Catalog No. H033425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H033425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H033425G Supplier TCI America Supplier No. H033425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used for research applications.

Chemical Identifiers

CAS 1191-43-1
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00004910
InChI Key SRZXCOWFGPICGA-UHFFFAOYSA-N
Synonym 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
PubChem CID 14491
IUPAC Name hexane-1,6-dithiol
SMILES SCCCCCCS

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 g
Formula Weight 150.30
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,6-Hexanedithiol
RTECSNumber MO3500000
TSCA No