missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich 1,6-Hexanediol
Description
1,6-Hexanediol
Specifications
Specifications
| CAS | 629-11-8 |
| Melting Point | 38°C to 42°C (lit.) |
| Boiling Point | 250°C (lit.) |
| Molecular Formula | C6H14O2 |
| Linear Formula | HO(CH2)6 OH |
| MDL Number | MFCD00002985 |
| Quantity | 2.5 kg |
| Synonym | Hexamethylene glycol |
| Molecular Weight (g/mol) | 118.17 |
| Percent Purity | 97% |