missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 98%, Spectrum™ Chemical

Catalog No. p-7165696
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
15 kg
Packaging:
Amber Glass Bottle
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608073 500 g Amber Glass Bottle
18608069 1 kg Amber Glass Bottle
18608070 5 kg Steel Pail
18608071 10 kg Steel Pail
18608072 15 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608073 Supplier Spectrum Chemical Mfg Cor Supplier No. HE170500GM
Only null left

Chemical Identifiers

CAS 124-09-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.