missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 98%, Spectrum™ Chemical

Catalog No. 18608073
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
15 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608073 500 g Amber Glass Bottle
18608069 1 kg Amber Glass Bottle
18608070 5 kg Amber Glass Bottle
18608071 10 kg Amber Glass Bottle
18608072 15 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608073 Supplier Spectrum Chemical Mfg Cor Supplier No. HE170500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-09-4

Specifications

CAS Max % 1
Boiling Point 204°C/399.2°F
Packaging Amber Glass Bottle
Quantity 500 g
Molecular Weight (g/mol) 116.2
Percent Purity 0.98
Grade Reagent
Physical Form Powder or Waxy Solid
Chemical Name or Material 1,6-Hexanediamine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.