missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 60 wt % aqueous soln.

Catalog No. AC411562500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411562500 250 g Plastic bottle
AC411560010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411562500 Supplier Thermo Scientific Chemicals Supplier No. 411562500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-09-4 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN

Specifications

Density 0.9300g/mL
Flash Point 116°C
Packaging Plastic bottle
Linear Formula NH2(CH2)6NH2
Quantity 250 g
Beilstein 04, 269
Specific Gravity 0.93
Merck Index 15, 4727
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Formula Weight 116.21
Physical Form Solution
Chemical Name or Material 1, 6-Hexanediamine, 60 wt % aqueous soln.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wate
EINECSNumber 204-679-6
RTECSNumber MO1180000
TSCA TSCA

RUO – Research Use Only