missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadiyne, 97%

CAS: 2396-63-6 | C7H8 | 92.141 g/mol

$210.07 - $2865.10

Chemical Identifiers

CAS 2396-63-6
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00014925
InChI Key RSPZSDWVQWRAEF-UHFFFAOYSA-N
Synonym 1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
PubChem CID 337121
IUPAC Name hepta-1,6-diyne
SMILES C#CCCCC#C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1131806
View Documents
Thermo Scientific Chemicals
A1131806
5 g
Each for $210.07
Only null left
 
AAA1131814
View Documents
Thermo Scientific Chemicals
A1131814
25 g
Each for $903.84
Only null left
 
AAA1131822
View Documents
Thermo Scientific Chemicals
A1131822
100 g
Each for $2,865.10
Only null left
 
Description

Description

1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Haevy metal powder, heat. Refrigerate at (+4°C).
Specifications

Chemical Identifiers

2396-63-6
92.141
RSPZSDWVQWRAEF-UHFFFAOYSA-N
337121
C#CCCCC#C
C7H8
MFCD00014925
1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
hepta-1,6-diyne

Specifications

2396-63-6
109°C to 111°C
Recognizable
1.442
5 g
1731756
Insoluble in water.
C#CCCCC#C
92.141
92.14
1,6-Heptadiyne
0.81
9°C (48°F)
C7H8
MFCD00014925
UN3295
1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
RSPZSDWVQWRAEF-UHFFFAOYSA-N
hepta-1,6-diyne
337121
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 219-253-5

RTECSNumber : MI5600000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only