missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dimethoxynaphthalene, 98+%

Catalog No. p-7047228
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1812903 1 g
AAL1812906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1812903 Supplier Thermo Scientific Chemicals Supplier No. L1812903
Only null left

CAS: 3900-49-0 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3900-49-0
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00086330
InChI Key RBUFUWIWCCOVOS-UHFFFAOYSA-N
Synonym naphthalene, 1,6-dimethoxy, 1,6-dimethoxy naphthalene, 1,6-dimethoxy-naphthalene, 1,6-dimethoxynaphtalene, 1,6-dimethoxynapthalene, maybridge3_002840, acmc-1bn8y, 1,6-dimethoxyl naphthalene, 1,6-dimethoxynaphthalene
PubChem CID 296916
IUPAC Name 1,6-dimethoxynaphthalene
SMILES COC1=CC=C2C(OC)=CC=CC2=C1

Specifications

Melting Point 58°C to 59°C
Boiling Point 123°C (0.8 mmHg)
Quantity 1 g
Beilstein 1945987
Formula Weight 188.23
Percent Purity ≥98%
Chemical Name or Material 1,6-Dimethoxynaphthalene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only