missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diisocyanatohexane, 99+%

Catalog No. p-113246
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115190250 25 g Glass bottle
AC115191000 100 g Glass bottle
AC115190010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC115190250 Supplier Thermo Scientific Chemicals Supplier No. 115190250

CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -67°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 255.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Linear Formula OCN(CH2)6NCO
Quantity 25 g
Beilstein 04, II, 711
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether
Formula Weight 168.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 6-Diisocyanatohexane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hai
EINECSNumber 212-485-8
RTECSNumber MO1740000
TSCA TSCA

RUO – Research Use Only