missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diiodohexane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. D244325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D244325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D244325G Supplier TCI America Supplier No. D244325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-09-4
Molecular Formula C6H12I2
Molecular Weight (g/mol) 337.97
MDL Number MFCD00001103
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
PubChem CID 12373
IUPAC Name 1,6-diiodohexane
SMILES ICCCCCCI

Specifications

Boiling Point 142°C
Quantity 25 g
Formula Weight 337.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Diiodohexane (stabilized with Copper chip)
RTECSNumber MO1700000
TSCA Yes