missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diiodohexane, 97+%, stab. with copper

Catalog No. AAA1536822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1536822 100 g
AAA1536814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1536822 Supplier Thermo Scientific Chemicals Supplier No. A1536822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-09-4 | C6H12I2 | 337.97 g/mol

1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 629-09-4
Molecular Formula C6H12I2
Molecular Weight (g/mol) 337.97
MDL Number MFCD00001103
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
PubChem CID 12373
IUPAC Name 1,6-diiodohexane
SMILES ICCCCCCI

Specifications

Density 2.052
Boiling Point 123°C to 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5852
Quantity 100 g
Beilstein 1280128
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 337.97
Percent Purity 98%
Chemical Name or Material 1,6-Diiodohexane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-073-5
RTECSNumber MO1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only