missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dicyanohexane, 99%

Catalog No. AC113861000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113861000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC113861000 Supplier Thermo Scientific Chemicals Supplier No. 113861000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-40-3 | C8H12N2 | 136.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-40-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00001981
InChI Key BTNXBLUGMAMSSH-UHFFFAOYSA-N
Synonym 1,6-dicyanohexane, suberonitrile, suberic acid dinitrile, suberodinitrile, unii-jnt1lxa02u, korksaeuredinitril german, jnt1lxa02u, hexamethylene di-isocyanide, hexane-1,6-dicarbonitrile, hexamethylenediisocyanic acid
PubChem CID 12385
IUPAC Name octanedinitrile
SMILES C(CCCC#N)CCC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -3.5°C
Color Yellow
Density 0.9500g/mL
Boiling Point 175.0°C (10.0 mmHg)
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450
Linear Formula NC(CH2)6CN
Quantity 100 mL
Beilstein 02, 694
Specific Gravity 0.95
Solubility Information Solubility in water: 1.2%
Viscosity 8.5 mPa.s (20°C)
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 6-Dicyanohexane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 211-089-2

RUO – Research Use Only