missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane 99.0+%, TCI America™

Catalog No. D009525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009525G 25 g
D0095500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D009525G Supplier TCI America Supplier No. D009525G

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN

Specifications

Melting Point 42°C
Color White
Boiling Point 205°C
Quantity 25 g
UN Number 2280
Formula Weight 116.21
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,6-Diaminohexane
EINECSNumber (2)-0153
RTECSNumber MO1180000
TSCA Yes