missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane Dihydrochloride 98.0+%, TCI America™

Catalog No. D0096500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0096500G 500 g
D009625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0096500G Supplier TCI America Supplier No. D0096500G

Chemical Identifiers

CAS 6055-52-3
Molecular Formula C6H18Cl2N2
Molecular Weight (g/mol) 189.124
MDL Number MFCD00012528
InChI Key XMVQMBLTFKAIOX-UHFFFAOYSA-N
Synonym 1,6-diaminohexane dihydrochloride, hexane-1,6-diamine dihydrochloride, 1,6-hexanediamine dihydrochloride, hexamethylenediamine dihydrochloride, 1,6-hexanediamine, dihydrochloride, unii-6if950zp7c, 1,6-hexanediamine 2hcl, hexanediamine dihydrochloride, hexamethylenediammonium dichloride, 1,6-hexamethylenediamine dihydrochloride
PubChem CID 2786615
IUPAC Name hexane-1,6-diamine;dihydrochloride
SMILES C(CCCN)CCN.Cl.Cl

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 500 g
Formula Weight 189.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Diaminohexane Dihydrochloride
EINECSNumber (2)-0153
RTECSNumber MO1312000
TSCA Yes