Learn More
1,6-Diaminohexane, 98+%
CAS: 124-09-4 | C6H16N2 | 116.208 g/mol
Supplier: Thermo Scientific Chemicals A1421222
Description
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.
Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.
Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.
Specifications
| 1,6-Diaminohexane | |
| 39°C to 43°C | |
| 12.4 | |
| 85°C (185°F) | |
| C6H16N2 | |
| MFCD00008243 | |
| UN2280 | |
| Air sensitive; Hygroscopic | |
| 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution | |
| NAQMVNRVTILPCV-UHFFFAOYSA-N | |
| hexane-1,6-diamine | |
| 16402 | |
| 116.21 |
| 124-09-4 | |
| 0.84 | |
| 199°C to 204°C | |
| Amine-like | |
| H2N(CH2)6NH2 | |
| 100 g | |
| 1098307 | |
| 14,4695 | |
| Soluble in water. Slightly soluble in ether,alcohol and benzene. | |
| C(CCCN)CCN | |
| 116.208 | |
| CHEBI:39618 | |
| ≥98% |
Chemical Identifiers
| 124-09-4 | |
| MFCD00008243 | |
| 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution | |
| CHEBI:39618 |
| C6H16N2 | |
| NAQMVNRVTILPCV-UHFFFAOYSA-N | |
| 16402 | |
| hexane-1,6-diamine |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HEXAMETHYLENEDIAMINE, SOLID
EINECSNumber : 204-679-6
RTECSNumber : MO1180000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only