missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane, 98+%

Catalog No. AAA1421236
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1421236 500 g
AAA1421222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1421236 Supplier Thermo Scientific Chemicals Supplier No. A1421236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-09-4 | C6H16N2 | 116.208 g/mol

1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine

Specifications

Melting Point 39°C to 43°C
Density 0.84
pH 12.4
Boiling Point 199°C to 204°C
Flash Point 85°C (185°F)
Odor Amine-like
Linear Formula H2N(CH2)6NH2
Quantity 500 g
UN Number UN2280
Beilstein 1098307
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,4695
Solubility Information Soluble in water. Slightly soluble in ether,alcohol and benzene.
SMILES C(CCCN)CCN
Molecular Weight (g/mol) 116.208
Formula Weight 116.21
Percent Purity ≥98%
Chemical Name or Material 1,6-Diaminohexane
Show More Show Less
Hazard Category H227-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HEXAMETHYLENEDIAMINE, SOLID
EINECSNumber 204-679-6
RTECSNumber MO1180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only