missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diacetoxyhexane 98.0+%, TCI America™

Catalog No. H0832500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0832500ML 500 mL
H083225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0832500ML Supplier TCI America Supplier No. H0832500ML

Chemical Identifiers

CAS 6222-17-9
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00191655
InChI Key ZMFWEWMHABZQNB-UHFFFAOYSA-N
Synonym 1,6-diacetoxyhexane, 1,6-hexanediol diacetate, hexylene glycol diacetate, 1,6-hexanediol, diacetate, 1,6-hexanediol, 1,6-diacetate, 1,6-dihydroxyhexane diacetate, hexamethylene diacetate, hexane-1,6-diyl diacetate, acmc-209n0s, 6-acetyloxy hexyl acetate
PubChem CID 80356
IUPAC Name 6-acetyloxyhexyl acetate
SMILES CC(=O)OCCCCCCOC(=O)C

Specifications

Color Yellow
Boiling Point 260°C
Quantity 500 mL
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Diacetoxyhexane
EINECSNumber (2)-3706
RTECSNumber MO2325000
TSCA Yes